4-Propylcyklohexanon ≥98.0%

Leverantör: TCI
Varning

P1849-25G P1849-5G
TCIAP1849-25GEA 775 SEK
TCIAP1849-25G TCIAP1849-5G
4-Propylcyklohexanon ≥98.0%
4-Propylcyklohexanon
Formel: C₉H₁₆O
Kokpunkt: 98°C
Densitet: 0,91 g/cm³ (20 °C)
Flampunkt: 86°C
MDL Number: MFCD00058690
CAS nummer: 40649-36-3
EINECS: 406-810-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93c0467297c74cc829dbc72bd69aa1868b9c6dce&vid=93c0467297c74cc829dbc72bd69aa1868b9c6dce&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR