3-Benzoylpropionic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid
B0868-25G
B0868-500G
B0868-100G
TCIAB0868-25GEA
485
SEK
TCIAB0868-25G
TCIAB0868-500G
TCIAB0868-100G
3-Benzoylpropionic acid ≥98.0% (by GC, titration analysis)
3-Benzoylpropionic acid
Formel:
C₁₀H₁₀O₃ MW: 178,19 g/mol Kokpunkt: 325 °C (1013 hPa) Smältpunkt: 114…117 °C |
MDL Number:
MFCD00002792 CAS nummer: 2051-95-8 EINECS: 218-135-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...