4-Formylbenzoic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Terephthalaldehydic acid, 4-Carboxybenzaldehyde
T0011-25G
T0011-500G
T0011-100G
TCIAT0011-25GEA
535
SEK
TCIAT0011-25G
TCIAT0011-500G
TCIAT0011-100G
4-Formylbenzoic acid ≥98.0%
4-Formylbenzoic acid
Formel:
C₈H₆O₃ MW: 150,13 g/mol Kokpunkt: 306 °C (1013 hPa) Smältpunkt: 256°C |
MDL Number:
MFCD00006951 CAS nummer: 619-66-9 EINECS: 210-607-4 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...