(±)-γ-Valerolactone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

V0007-25G V0007-500G
TCIAV0007-25GEA 215 SEK
TCIAV0007-25G TCIAV0007-500G
(±)-γ-Valerolactone ≥98.0%
(±)-γ-Valerolactone
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 207°C
Smältpunkt: -31°C
Densitet: 1,05 g/cm³ (20 °C)
Flampunkt: 102°C
MDL Number: MFCD00005400
CAS nummer: 108-29-2
EINECS: 203-569-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f7f4f84778c5e747e496738c35ec49303ea775&vid=f8f7f4f84778c5e747e496738c35ec49303ea775&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR