6-Methyl-2-thiouracil ≥98.0%

Leverantör: TCI
Varning

M0443-25G M0443-500G
TCIAM0443-25GEA 245 SEK
TCIAM0443-25G TCIAM0443-500G
6-Methyl-2-thiouracil ≥98.0%
6-Methyl-2-thiouracil
Formel: C₅H₆N₂OS
Kokpunkt: 351 °C (1013 hPa)
Densitet: 1,519 g/cm³ (20 °C)
MDL Number: MFCD00006040
CAS nummer: 56-04-2
EINECS: 200-252-3
Merck Index: 14,06128
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52921b5c2bd6e60052d0e8a600cd83f7a9e9ec68&vid=52921b5c2bd6e60052d0e8a600cd83f7a9e9ec68&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR