5-Methyl-2-thiouracil ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 4-Hydroxy-2-mercapto-5-methylpyrimidine, 2-Mercapto-5-methylpyrimidin-4-ol, 4-Hydroxy-5-methyl-2-thiopyrimidine
M0994-10G M0994-25G
TCIAM0994-10GEA 1100 SEK
TCIAM0994-10G TCIAM0994-25G
5-Methyl-2-thiouracil ≥98.0%
5-Methyl-2-thiouracil
Formel: C₅H₆N₂OS
MW: 142,18 g/mol
Kokpunkt: 367 °C (1013 hPa)
Smältpunkt: 265 °C
Densitet: 1,49 g/cm³ (22 °C)
MDL Number: MFCD00009777
CAS nummer: 636-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79b2680dbde22b07083f2af1500160f88c0ff5f1&vid=79b2680dbde22b07083f2af1500160f88c0ff5f1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency