4,5-Dimethylthiazole ≥98.0%

Leverantör: TCI
Varning

D2142-25G D2142-5G
TCIAD2142-25GEA 830 SEK
TCIAD2142-25G TCIAD2142-5G
4,5-Dimethylthiazole ≥98.0%
4,5-Dimethylthiazole
Formel: C₅H₇NS
Kokpunkt: 159°C
Densitet: 1,0828 g/cm³ (20 °C)
Flampunkt: 51°C
MDL Number: MFCD00005336
CAS nummer: 3581-91-7
EINECS: 222-703-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12c4ec9183944c20d610dd5fe01b1407a7c1cb36&vid=12c4ec9183944c20d610dd5fe01b1407a7c1cb36&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR