3-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Varning
|
N0243-25G
N0243-500G
TCIAN0243-25GEA
215
SEK
TCIAN0243-25G
TCIAN0243-500G
3-Nitrophthalic acid ≥98.0% (by HPLC, titration analysis)
3-Nitrophthalic acid
Formel:
C₈H₅NO₆ Kokpunkt: 441 °C (1013 hPa) Smältpunkt: 210…215 °C |
MDL Number:
MFCD00007138 CAS nummer: 603-11-2 EINECS: 210-030-8 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...