Methyl-m-toluate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: m-Toluic acid methyl ester, Methyl 3-methylbenzoate, 3-Toluic acid methyl ester, Methyl-3-methylbenzoate, 3-Methylbenzoic acid methyl ester

T1258-25ML T1258-500ML
TCIAT1258-25MLEA 325 SEK
TCIAT1258-25ML TCIAT1258-500ML
Methyl-m-toluate ≥98.0%
Methyl-m-toluate
Formel: C₉H₁₀O₂
MW: 150,18 g/mol
Kokpunkt: 115°C
Flampunkt: 96°C
MDL Number: MFCD00008436
CAS nummer: 99-36-5
EINECS: 202-753-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f45e2650910b66ab9c0db5d6d946bd9a7e1feb0&vid=5f45e2650910b66ab9c0db5d6d946bd9a7e1feb0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR