3-Fluorophthalic anhydride ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
4-Fluoroisobenzofuran-1,3-dione
F0349-25G
F0349-5G
TCIAF0349-25GEA
5375
SEK
TCIAF0349-25G
TCIAF0349-5G
3-Fluorophthalic anhydride ≥98.0%
3-Fluorophthalic anhydride
Formel:
C₈H₃FO₃ Kokpunkt: 283°C Smältpunkt: 162…163 °C |
MDL Number:
MFCD00039696 CAS nummer: 652-39-1 EINECS: 211-491-8 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Morpholine Method) | min. 97.0 % |
Melting point | 160.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...