5-Amino-2-methoxypyridine ≥97.0% (by GC, titration analysis)
Leverantör: TCI
Varning
A1956-25G A1956-5G
TCIAA1956-25GEA 1750 SEK
TCIAA1956-25G TCIAA1956-5G
5-Amino-2-methoxypyridine ≥97.0% (by GC, titration analysis)
5-Amino-2-methoxypyridine
Formel: C₆H₈N₂O
MW: 124.14 g/mol
Kokpunkt: 120°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006264
CAS nummer: 6628-77-9
EINECS: 229-612-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=554da6a715511426d6ac0822be7e55a4e8d83530&vid=554da6a715511426d6ac0822be7e55a4e8d83530&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %