3,5-Dichlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,5-Dichloro-2-hydroxybenzaldehyde
D1170-25G
D1170-5G
TCIAD1170-25GEA
1075
SEK
TCIAD1170-25G
TCIAD1170-5G
3,5-Dichlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
3,5-Dichlorosalicylaldehyde
Formel:
C₇H₄Cl₂O₂ MW: 191,01 g/mol Kokpunkt: 723 °C (1013 hPa) Smältpunkt: 92 °C |
MDL Number:
MFCD00003320 CAS nummer: 90-60-8 EINECS: 202-005-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Melting point | 93.0 to 96.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...