3-Thiopheneethanol ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol

T1475-25G T1475-5G
TCIAT1475-25GEA 2275 SEK
TCIAT1475-25G TCIAT1475-5G
3-Thiopheneethanol ≥97.0%
3-Thiopheneethanol
Formel: C₆H₈OS
MW: 128,19 g/mol
Kokpunkt: 111°C
Densitet: 1,14 g/cm³ (20 °C)
Flampunkt: 92°C
MDL Number: MFCD00009766
CAS nummer: 13781-67-4
EINECS: 237-434-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c4a4c0945d08e1c33000840b9b0325c0dcfb39c&vid=6c4a4c0945d08e1c33000840b9b0325c0dcfb39c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR