2-Thenoic acid ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid
T0218-25G T0218-100G
TCIAT0218-25GEA 485 SEK
TCIAT0218-25G TCIAT0218-100G
2-Thenoic acid ≥98.0%
2-Thenoic acid
Formel: C₅H₄O₂S
MW: 128,15 g/mol
Kokpunkt: 260 °C (1013 hPa)
Smältpunkt: 123…125 °C
MDL Number: MFCD00005437
CAS nummer: 527-72-0
EINECS: 208-423-4
Merck Index: 14,09354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33de5a2074a6b065870b7a5db77225ca755a9eb2&vid=33de5a2074a6b065870b7a5db77225ca755a9eb2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 130.0 deg-C