2-Methylfuran ≥98.0%

Leverantör: TCI
Fara

M0226-25ML M0226-500ML
TCIAM0226-25MLEA 215 SEK
TCIAM0226-25ML TCIAM0226-500ML
2-Methylfuran ≥98.0%
2-Methylfuran
Formel: C₅H₆O
MW: 82.1 g/mol
Kokpunkt: 63°C
Smältpunkt: -89
Flampunkt: –30°C
MDL Number: MFCD00003248
CAS nummer: 534-22-5
EINECS: 208-594-5
UN: 2301
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb3454d61ac4a17e756c42024e749ea43f184a5&vid=ebb3454d61ac4a17e756c42024e749ea43f184a5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR