2-Methylbenzothiazole ≥98.0%
Leverantör: TCI
M0516-25G
M0516-500G
M0516-100G
TCIAM0516-25GEA
1225
SEK
TCIAM0516-25G
TCIAM0516-500G
TCIAM0516-100G
2-Methylbenzothiazole ≥98.0%
2-Methylbenzothiazole
Formel:
C₈H₇NS MW: 149.22 g/mol Kokpunkt: 238°C Smältpunkt: 12 Flampunkt: 123°C |
MDL Number:
MFCD00005794 CAS nummer: 120-75-2 EINECS: 204-423-3 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...