Benzotiazol-2-tiol ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
M0055-25G M0055-500G
TCIAM0055-25GEA 215 SEK
TCIAM0055-25G TCIAM0055-500G
Benzotiazol-2-tiol ≥98.0% (by HPLC, titration analysis)
Benzotiazol-2-tiol
Formel: C₇H₅NS₂
MW: 167,26 g/mol
Kokpunkt: 302 °C (1013 hPa)
Smältpunkt: 177…182 °C
Densitet: 1,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005781
CAS nummer: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 14,05868
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36bae1e7599a9a34669875ab9fbf318a4b71a4fa&vid=36bae1e7599a9a34669875ab9fbf318a4b71a4fa&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Green to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 183.0 deg-C