2-Ethyl-4-methylthiazole ≥98.0%

Leverantör: TCI
Varning

E0456-25G E0456-5G
TCIAE0456-25GEA 1225 SEK
TCIAE0456-25G TCIAE0456-5G
2-Ethyl-4-methylthiazole ≥98.0%
2-Ethyl-4-methylthiazole
Formel: C₆H₉NS
Kokpunkt: 67°C
Densitet: 1,0231 g/cm³ (25 °C)
Flampunkt: 54°C
MDL Number: MFCD00036621
CAS nummer: 15679-12-6
EINECS: 239-757-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ced2a30f6fdc7e0f7f32dd10196c20203ded79&vid=06ced2a30f6fdc7e0f7f32dd10196c20203ded79&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR