2-Acetylthiazole ≥98.0%

Leverantör: TCI
Varning

A1265-25G A1265-5G
TCIAA1265-25GEA 3200 SEK
TCIAA1265-25G TCIAA1265-5G
2-Acetylthiazole ≥98.0%
2-Acetylthiazole
Formel: C₅H₅NOS
Kokpunkt: 90°C
Densitet: 1,22 g/cm³ (20 °C)
Flampunkt: 78°C
MDL Number: MFCD00005324
CAS nummer: 24295-03-2
EINECS: 246-134-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0eb830fdc1d699bfc6b934db562d15cf29038a0&vid=c0eb830fdc1d699bfc6b934db562d15cf29038a0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR