2-Chlorothiazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C2471-25G
C2471-5G
TCIAC2471-25GEA
2250
SEK
TCIAC2471-25G
TCIAC2471-5G
2-Chlorothiazole ≥98.0%
2-Chlorothiazole
Formel:
C₃H₂ClNS MW: 119,57 g/mol Kokpunkt: 145°C Flampunkt: 54°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00210701 CAS nummer: 3034-52-4 EINECS: 221-228-9 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...