2-Chlorobenzoxazole ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
2-Chloro-1,3-benzoxazole
C1387-25G
C1387-5G
TCIAC1387-25GEA
1325
SEK
TCIAC1387-25G
TCIAC1387-5G
2-Chlorobenzoxazole ≥98.0%
2-Chlorobenzoxazole
Formel:
C₇H₄ClNO MW: 153.57 g/mol Kokpunkt: 202°C Smältpunkt: 7°C Flampunkt: 85°C |
MDL Number:
MFCD00005766 CAS nummer: 615-18-9 EINECS: 210-414-5 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...