2-Acetyl-5-bromothiophene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(5-Bromo-2-thienyl)-1-ethanone
A1183-25G
A1183-5G
TCIAA1183-25GEA
950
SEK
TCIAA1183-25G
TCIAA1183-5G
2-Acetyl-5-bromothiophene ≥98.0%
2-Acetyl-5-bromothiophene
Formel:
C₆H₅BrOS MW: 205,08 g/mol Kokpunkt: 103°C Smältpunkt: 92…93 °C |
MDL Number:
MFCD00014528 CAS nummer: 5370-25-2 EINECS: 226-363-7 |
Specifikation testresultat
Appearance | White to Brown powder to crystalline |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...