2-Aminothiazole ≥98.0%

Leverantör: TCI
Varning

A0633-25G A0633-500G A0633-100G
TCIAA0633-25GEA 215 SEK
TCIAA0633-25G TCIAA0633-500G TCIAA0633-100G
2-Aminothiazole ≥98.0%
2-Aminothiazole
Formel: C₃H₄N₂S
Kokpunkt: 143°C
Smältpunkt: 86…89 °C
Densitet: 1,5 g/cm³ (20 °C)
MDL Number: MFCD00005325
CAS nummer: 96-50-4
EINECS: 202-511-6
Merck Index: 14,00479
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c11184de61875cff6660d0c56c971bb26c80b53&vid=6c11184de61875cff6660d0c56c971bb26c80b53&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR