2-Amino-6-methylbenzothiazole ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Varning
Synonyms: 6-Methylbenzothiazol-2-ylamine
A0714-100G A0714-25G
TCIAA0714-100GEA 2400 SEK
TCIAA0714-100G TCIAA0714-25G
2-Amino-6-methylbenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-6-methylbenzothiazole
Formel: C₈H₈N₂S
MW: 164.23 g/mol
Kokpunkt: 321
Smältpunkt: 142
MDL Number: MFCD00005789
CAS nummer: 2536-91-6
EINECS: 219-801-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e78e8b7d19a2afcb9a23b76808334d900b6b6a&vid=15e78e8b7d19a2afcb9a23b76808334d900b6b6a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 141.0 deg-C