2-Amino-6-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 6-Chlorobenzothiazol-2-ylamine, 6-Chloro-1,3-benzothiazol-2-amine

A1216-25G A1216-500G
TCIAA1216-25GEA 325 SEK
TCIAA1216-25G TCIAA1216-500G
2-Amino-6-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chlorobenzothiazole
Formel: C₇H₅ClN₂S
MW: 184.65 g/mol
Kokpunkt: 344
MDL Number: MFCD00053557
CAS nummer: 95-24-9
EINECS: 202-402-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40945141e60bdb671bc9ef662388b3de98d4fbee&vid=40945141e60bdb671bc9ef662388b3de98d4fbee&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR