2-Amino-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine

A0381-25G A0381-500G
TCIAA0381-25GEA 965 SEK
TCIAA0381-25G TCIAA0381-500G
2-Amino-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-nitrothiazole
Formel: C₃H₃N₃O₂S
MW: 145,14 g/mol
Smältpunkt: 203 °C
MDL Number: MFCD00005326
CAS nummer: 121-66-4
EINECS: 204-490-9
Merck Index: 14,00457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62b101690e026e8444c42dfe707fc3e91028b581&vid=62b101690e026e8444c42dfe707fc3e91028b581&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR