2-Amino-2-ethyl-1,3-propanediol ≥98.0%

Leverantör: TCI
Varning

A0620-25G A0620-500G
TCIAA0620-25GEA 215 SEK
TCIAA0620-25G TCIAA0620-500G
2-Amino-2-ethyl-1,3-propanediol ≥98.0%
2-Amino-2-ethyl-1,3-propanediol
Formel: C₅H₁₃NO₂
Kokpunkt: 152°C
Smältpunkt: 39 °C
Densitet: 1,099 g/cm³ (20 °C)
MDL Number: MFCD00004680
CAS nummer: 115-70-8
EINECS: 204-101-2
Merck Index: 14,00439
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45da71b5180e4a9118851c6e27bd0acd0efd3ffc&vid=45da71b5180e4a9118851c6e27bd0acd0efd3ffc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange lump
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR