2,6-Dichlorobenzothiazole ≥98.0%
Leverantör: TCI EUROPE
Varning
D3801-1G D3801-5G
TCIAD3801-1GEA 725 SEK
TCIAD3801-1G TCIAD3801-5G
2,6-Dichlorobenzothiazole ≥98.0%
2,6-Dichlorobenzothiazole
Formel: C₇H₃Cl₂NS
MW: 204,08 g/mol
MDL Number: MFCD00044101
CAS nummer: 3622-23-9
EINECS: 222-819-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55bd96a775c545604329199637f3a5334e8040df&vid=55bd96a775c545604329199637f3a5334e8040df&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C