Pentaerytritol ≥98.0%

Leverantör: TCI EUROPE

P0039-500G P0039-25G
TCIAP0039-500GEA 310 SEK
TCIAP0039-500G TCIAP0039-25G
Pentaerytritol ≥98.0%
Pentaerytritol
Formel: C₅H₁₂O₄
MW: 136,15 g/mol
Kokpunkt: 276°C
Smältpunkt: 260…264 °C
Densitet: 1,511 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004692
CAS nummer: 115-77-5
EINECS: 204-104-9
Merck Index: 14,07111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1013feab1205250834f780be1c8a5ae0a310985&vid=f1013feab1205250834f780be1c8a5ae0a310985&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR