Furfurylalkohol ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Furfurol

F0076-25ML F0076-500ML
TCIAF0076-25MLEA 230 SEK
TCIAF0076-25ML TCIAF0076-500ML
Furfurylalkohol ≥98.0%
Furfurylalkohol
Formel: C₅H₆O₂
MW: 98,1 g/mol
Kokpunkt: 170°C
Smältpunkt: –31 °C
Densitet: 1,13 g/cm³ (20 °C)
Flampunkt: 78°C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003252
CAS nummer: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 14,04305
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a189781463810bf1b4d203b0d940e54a34d2de56&vid=a189781463810bf1b4d203b0d940e54a34d2de56&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR