α,α,α-Trifluoro-o-xylene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Methylbenzotrifluoride, 1-Methyl-2-(trifluoromethyl)benzene, 2-(Trifluoromethyl)toulene

M1215-1G M1215-5G
TCIAM1215-1GEA 495 SEK
TCIAM1215-1G TCIAM1215-5G
α,α,α-Trifluoro-o-xylene ≥98.0%
α,α,α-Trifluoro-o-xylene
Formel: C₈H₇F₃
MW: 160,14 g/mol
Kokpunkt: 126 °C (1013 hPa)
Densitet: 1,174 g/cm³ (19 °C)
MDL Number: MFCD00153204
CAS nummer: 13630-19-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44e7a369ce51cb38cc0739c8fe3862f7b8c0c174&vid=44e7a369ce51cb38cc0739c8fe3862f7b8c0c174&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR