4'-Hydroxyvalerophenone ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
1-(4-Hydroxyphenyl)-1-pentanone
H0627-25G
H0627-500G
TCIAH0627-25GEA
1025
SEK
TCIAH0627-25G
TCIAH0627-500G
4'-Hydroxyvalerophenone ≥98.0%
4'-Hydroxyvalerophenone
Formel:
C₁₁H₁₄O₂ MW: 178.23 g/mol Smältpunkt: 77 |
MDL Number:
MFCD00009719 CAS nummer: 2589-71-1 EINECS: 219-978-7 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 64.0 to 67.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...