1-Ethyl-3-methylimidazolium ethylsulphate ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
EMIM ESU
E0650-25G
E0650-5G
TCIAE0650-25GEA
965
SEK
TCIAE0650-25G
TCIAE0650-5G
1-Ethyl-3-methylimidazolium ethylsulphate ≥98.0%
1-Ethyl-3-methylimidazolium ethylsulphate
Formel:
C₈H₁₆N₂O₄S Flampunkt: 162°C |
MDL Number:
MFCD06798189 CAS nummer: 342573-75-5 |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...