Valeronitrile ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Pentanenitrile
V0008-25ML
V0008-500ML
TCIAV0008-25MLEA
215
SEK
TCIAV0008-25ML
TCIAV0008-500ML
Valeronitrile ≥98.0%
Valeronitrile
Formel:
C₅H₉N MW: 83.13 g/mol Kokpunkt: 140°C Smältpunkt: –96°C Flampunkt: 40°C |
MDL Number:
MFCD00001974 CAS nummer: 110-59-8 EINECS: 203-781-8 UN: 3273 ADR: 3 II Merck Index: 14,09905 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...