α,α,α-Trifluoro-p-toluonitrile ≥98.0%
Leverantör: TCI
Fara
Synonyms: 4-(Trifluoromethyl)benzonitrile, 4-Cyanobenzotrifluoride, p-(Trifluoromethyl)benzonitrile
C1005-25G C1005-5G
TCIAC1005-25GEA 670 SEK
TCIAC1005-25G TCIAC1005-5G
α,α,α-Trifluoro-p-toluonitrile ≥98.0%
α,α,α-Trifluoro-p-toluonitrile
Formel: C₈H₄F₃N
Kokpunkt: 81°C
Smältpunkt: 35…38 °C
Densitet: 1,2784 g/cm³ (20 °C)
Flampunkt: (closed cup)
MDL Number: MFCD00001826
CAS nummer: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f6a78afe7a9f1d8c56c738f4dcacdad94bd7eb3&vid=9f6a78afe7a9f1d8c56c738f4dcacdad94bd7eb3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %