5-Chlorovaleronitrile ≥96.0%

Leverantör: TCI
Fara

Synonyms: 5-Chloropentanenitrile

C1421-25G C1421-250G
TCIAC1421-25GEA 550 SEK
TCIAC1421-25G TCIAC1421-250G
5-Chlorovaleronitrile ≥96.0%
5-Chlorovaleronitrile
Formel: C₅H₈ClN
MW: 117.58 g/mol
Kokpunkt: 118°C
Flampunkt: 98°C
MDL Number: MFCD00001977
CAS nummer: 6280-87-1
EINECS: 228-481-4
UN: 3276
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f6f34f6704cc60c620347b5bb871ef8385d8c7&vid=f0f6f34f6704cc60c620347b5bb871ef8385d8c7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR