4-Chloro-m-phenylenediamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

C0710-25G C0710-5G
TCIAC0710-25GEA 2250 SEK
TCIAC0710-25G TCIAC0710-5G
4-Chloro-m-phenylenediamine ≥98.0% (by GC, titration analysis)
4-Chloro-m-phenylenediamine
Formel: C₆H₇ClN₂
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 91 °C
MDL Number: MFCD00025284
CAS nummer: 5131-60-2
EINECS: 225-877-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ab183a6ffc66bf2fbec4388b07299642699eabf&vid=9ab183a6ffc66bf2fbec4388b07299642699eabf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 89.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR