2-Chlorobenzyl mercaptan ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
2-Chlorobenzenemethanethiol, o-Chlorotoluene-α-thiol
C2068-25G
C2068-5G
TCIAC2068-25GEA
815
SEK
TCIAC2068-25G
TCIAC2068-5G
2-Chlorobenzyl mercaptan ≥98.0%
2-Chlorobenzyl mercaptan
Formel:
C₇H₇ClS Kokpunkt: 121°C Densitet: 1,22 g/cm³ (20 °C) Flampunkt: 69°C |
MDL Number:
MFCD00004868 CAS nummer: 39718-00-8 EINECS: 254-602-5 UN: 2810 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...