4-Butylphenyl isocyanate ≥98.0%
Leverantör: TCI
Fara
B3205-1G B3205-5G
TCIAB3205-1GEA 285 SEK
TCIAB3205-1G TCIAB3205-5G
4-Butylphenyl isocyanate ≥98.0%
4-Butylphenyl isocyanate
Formel: C₁₁H₁₃NO
Kokpunkt: 170°C
Densitet: 0,992 g/cm³ (20 °C)
Flampunkt: 108°C
Storage Temperature: Kylskåp
MDL Number: MFCD00013882
CAS nummer: 69342-47-8
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9436f273e80b4a3ad5d85d4a9d5321362c0fb1b&vid=c9436f273e80b4a3ad5d85d4a9d5321362c0fb1b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %