1-Butyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Fara
Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]
B2195-25G B2195-5G B2195-100G
TCIAB2195-25GEA 750 SEK
TCIAB2195-25G TCIAB2195-5G TCIAB2195-100G
1-Butyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazoliumtetrafluoroborate
Formel: C₈H₁₅BF₄N₂
MW: 226,02 g/mol
Kokpunkt: ? 200 °C (1013 hPa)
Smältpunkt: –71 °C
Densitet: 1,2 g/cm³ (20 °C)
Flampunkt: ≥ 110 °C
MDL Number: MFCD03095449
CAS nummer: 174501-65-6
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d996eee924329e9811b44e65664826eb95094508&vid=d996eee924329e9811b44e65664826eb95094508&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %