4-Butoxyphenol ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Hydroquinone monobutyl ether

B0923-25G B0923-500G
TCIAB0923-25GEA 535 SEK
TCIAB0923-25G TCIAB0923-500G
4-Butoxyphenol ≥97.0%
4-Butoxyphenol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Kokpunkt: 165°C
Smältpunkt: 65…66 °C
MDL Number: MFCD00002336
CAS nummer: 122-94-1
EINECS: 204-583-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7ed1e0a9bae5fcfd9d435d32ac4f4b7f40cbdad&vid=e7ed1e0a9bae5fcfd9d435d32ac4f4b7f40cbdad&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 64.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR