4-Bromobutyronitrile ≥97.0%

Leverantör: TCI
Fara

B0565-25G B0565-5G
TCIAB0565-25GEA 855 SEK
TCIAB0565-25G TCIAB0565-5G
4-Bromobutyronitrile ≥97.0%
4-Bromobutyronitrile
Formel: BrH₂CCH₂CH₂CN
Kokpunkt: 205°C
Densitet: 1,49 g/cm³ (20 °C)
Flampunkt: 103°C
MDL Number: MFCD00001971
CAS nummer: 5332-06-9
EINECS: 226-236-6
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba9efdcfe6bb0b51362ae15eb3532074b29f82&vid=d0ba9efdcfe6bb0b51362ae15eb3532074b29f82&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR