N-Bromophthalimide ≥95.0%
Leverantör: TCI
|
Varning
|
B1697-25G
B1697-5G
TCIAB1697-25GEA
1275
SEK
TCIAB1697-25G
TCIAB1697-5G
N-Bromophthalimide ≥95.0%
N-Bromophthalimide
Formel:
C₈H₄BrNO₂ MW: 226.03 g/mol |
MDL Number:
MFCD00005888 CAS nummer: 2439-85-2 EINECS: 219-467-9 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Melting point | 204.0 to 210.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...