m-Kresidin ≥99.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
Synonyms:
4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole
M1474-25G
M1474-500G
TCIAM1474-25GEA
1025
SEK
TCIAM1474-25G
TCIAM1474-500G
m-Kresidin ≥99.0% (by GC, titration analysis)
m-Kresidin
Formel:
C₈H₁₁NO MW: 137.18 g/mol Kokpunkt: 249°C Smältpunkt: 13°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007735 CAS nummer: 102-50-1 EINECS: 203-036-7 UN: 2810 |
|
Specifikation testresultat
Appearance | White to Brown powder to lump |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...