p-Fenetedin ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 4-Etoxianilin

P0090-25G P0090-500G
TCIAP0090-25GEA 215 SEK
TCIAP0090-25G TCIAP0090-500G
p-Fenetedin ≥98.0%
p-Fenetedin
Formel: C₈H₁₁NO
MW: 137,18 g/mol
Kokpunkt: 254°C
Smältpunkt: 5°C
Densitet: 1,06 g/cm³ (20 °C)
Flampunkt: 116°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007865
CAS nummer: 156-43-4
EINECS: 205-855-5
UN: 2311
ADR: 6.1,III
Merck Index: 14,07228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=894deeea54f509581256cf871bda2326de07985f&vid=894deeea54f509581256cf871bda2326de07985f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR