1-Amino-3-methoxypropane ≥99.0%
Leverantör: TCI
|
Fara
|
M0127-25ML
M0127-500ML
TCIAM0127-25MLEA
230
SEK
TCIAM0127-25ML
TCIAM0127-500ML
1-Amino-3-methoxypropane ≥99.0%
1-Amino-3-methoxypropane
Formel:
C₄H₁₁NO Kokpunkt: 116°C Smältpunkt: < –70 °C Densitet: 0,8737 g/cm³ (20 °C) Flampunkt: 32°C |
MDL Number:
MFCD00014831 CAS nummer: 5332-73-0 EINECS: 226-241-3 UN: 2734 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...