5-Chloro-o-anisidine ≥98.0%

Leverantör: TCI
Fara

Synonyms: 5-Chloro-2-methoxyaniline, 5-Chloro-2-anisidine, 2-Amino-4-chloroanisole

C0118-25G C0118-500G
TCIAC0118-25GEA 455 SEK
TCIAC0118-25G TCIAC0118-500G
5-Chloro-o-anisidine ≥98.0%
5-Chloro-o-anisidine
Formel: C₇H₈ClNO
MW: 157.6 g/mol
Kokpunkt: 259
Smältpunkt: 84
MDL Number: MFCD00007777
CAS nummer: 95-03-4
EINECS: 202-385-2
UN: 2233
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f7b1ddb5cec9396fc92167ed44fdbb41586dec&vid=53f7b1ddb5cec9396fc92167ed44fdbb41586dec&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR