α,α,α',α'-Tetrafluoro-p-xylene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
α,α,β,β-Tetrafluoro-p-xylene, 1,4-Bis(difluoromethyl)benzene
T1774-25G
T1774-5G
TCIAT1774-25GEA
3450
SEK
TCIAT1774-25G
TCIAT1774-5G
α,α,α',α'-Tetrafluoro-p-xylene ≥98.0%
α,α,α',α'-Tetrafluoro-p-xylene
Formel:
C₈H₆F₄ MW: 178,13 g/mol Kokpunkt: 70°C |
MDL Number:
MFCD01320708 CAS nummer: 369-54-0 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...