3,5-Dibromoanisole ≥98.0%

Leverantör: TCI
Varning

D3364-25G D3364-5G
TCIAD3364-25GEA 1700 SEK
TCIAD3364-25G TCIAD3364-5G
3,5-Dibromoanisole ≥98.0%
3,5-Dibromoanisole
Formel: C₇H₆Br₂O
Kokpunkt: 261°C
MDL Number: MFCD02258848
CAS nummer: 74137-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e1963e65434cdf217175032fd2c7619a8cb7c3&vid=27e1963e65434cdf217175032fd2c7619a8cb7c3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR