1,3-Diacetylbenzene ≥98.0%

Leverantör: TCI

D2617-5G D2617-1G
TCIAD2617-5GEA 645 SEK
TCIAD2617-5G TCIAD2617-1G
1,3-Diacetylbenzene ≥98.0%
1,3-Diacetylbenzene
Formel: C₁₀H₁₀O₂
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 72…79 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008740
CAS nummer: 6781-42-6
EINECS: 229-842-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba9a44edb7a662672dd5c7a88bf4ead3a0489470&vid=ba9a44edb7a662672dd5c7a88bf4ead3a0489470&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR